Learn More
BRANSON ULTRASONICS CORPORATION (3P) DIGITAL ULTRASONIC BATH
Supplier:  BRANSON ULTRASONICS CORPORATION (3P) CPX952219R
Encompass_Preferred
Branson Ultrasonics CPX Series Ultrasonic Cleaning Bath, Model: CPX2800, Capacity: 2.8L (0.75 gal.), Dimensions Interior: 23 x 14 x 10cm (9.5 x 5.5 x 4 in.), Voltage: 120V, Hertz: 40kHz, For Use With: Cleaning, Includes: Tank cover, Plastic, Timer: 90 min., Warranty: 2 Years, Weight: 9 lb., 4.1kg
Catalog No. NC3721486