Learn More
Cayman Chemical DIgItls pururea DIgItIn 250mg
SDP
Supplier:  Cayman Chemical 14952250 MG
Encompass_Preferred
A non-ionic detergent used to solubilize membrane-bound proteins, to precipitate cholesterol, and to permeabilize cell membranes
Catalog No. 50-296-9518