Learn More
AG Scientific Inc Digitonin 500 MG
Supplier:  AG Scientific Inc D2425500MG
Encompass_Preferred
Digitonin is a steroid glycoside isolated from Digitalis purpurea. It belongs to the class of non-ionic detergents commonly used for solubilization of membrane proteins and permeabilization of cellular and nuclear membranes.Digitonin permeabilizes plasma membranes of eukaryotic cells by complexing with membrane cholesterol and other unconjugated ß-hydroxysterols.
Catalog No. 50-226-8285