Learn More
Sigma Aldrich Fine Chemicals Biosciences DigiTrol II temperature co1EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z2854981EA
Encompass
Simultaneously displays set point and bath temperature to 1.0C. Supplied with a Type K 6 in. immersion thermocouple probe that fits Aldrich oil baths. 240 V models are supplied with a US connector.
Catalog No. 50-290-8820