Learn More
Enzo Life Sciences Diglycyl Lysine monoclonal antibody (GX41) (100µg)
SDP
Supplier:  Enzo Life Sciences ADI9083100100
Encompass_Preferred
Highly specific monoclonal for ubiquitin remnant profiling via immunoprecipitation. Clone: GX41. Host: Mouse. Isotype: IgG1κ. Immunogen: Diglycine-modified histone from calf thymus, Type III-S, lysine rich fraction. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: Species independent. Specificity: Recognizes diglycyl modified lysine, a ubiquitin remnant detectable after trypsin digestion. Applications: IP, WB. Purity: Protein G purified. Formulation: Liquid. In PBS containing 0.1% sodium azide. Handling: Store unopened vial at -20°C until required for use. Store diluted antibody at 2-4°C and use within 1 month. Avoid freeze/thaw cycles.
Catalog No. 50-200-7101