Learn More
Cayman Chemical DIglycylLysIn MoncLNal n 100ug
SDP
Supplier:  Cayman Chemical 21096100 UG
Encompass_Preferred
Immunogen: Diglycine-modified histone peptide • Host: Mouse • Species Reactivity: Species independent • Cross Reactivity: (+) Diglycyl-modified lysine, (-) Unmodified lysine • Applications: IP and WB
Catalog No. 50-298-2344