Learn More
Sigma Aldrich Fine Chemicals Biosciences Digoxigenin 11 dUTP alkali
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 11573152910
Encompass
Digoxigenin (DIG)-11-deoxyuridine triphosphate (dUTP) alkali-labile is provided as in 1 mM tetralithium salt SOLUTIONon. In which DIG is bound to dUTP via an alkali-sensitive ester linkage.
Catalog No. 50-290-5021