Learn More
Selleck Chemical LLC Digoxin S4290-500mg
SDP
Supplier:  Selleck Chemical LLC S4290500MG
Encompass_Preferred
Digoxin is a classical Na K-ATPase inhibitor with selectivity for the 2 3 isoform over the common 1 1 isoform used in the treatment of atrial fibrillation and heart failure This product is a hazardous chemical (acute toxicity/flammable/skin corrosive) Please use it while wearing a protective face mask gloves and clothing
Catalog No. 50-311-0178