Learn More
Selleck Chemical LLC Dihexa E4606-1G
SDP
Supplier:  Selleck Chemical LLC E46061G
Encompass_Preferred
Dihexa(PNB-0408) an oligopeptide drug is an orally active and blood-brain barrier-permeable analog of angiotensin IV It binds to hepatocyte growth factor (HGF) with high affinity and potentiates its activity at its receptor c-Met Dihexa can also be used as a therapeutic potential in the treatment of Alzheimer s disease
Catalog No. 50-000-38921