Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihexyl phthalate | 84-75-3 | 98.0% | 334.46 | 1 G
SDP

Supplier:  Medchemexpress LLC HYW0112151G

Encompass_Preferred

Dihexyl phthalate is a commonly used phthalate ester found in various plastics and consumer products. It is classified as a priority pollutant and an endocrine disruptor, capable of inducing reactive oxygen species (ROS) accumulation and promoting inflammation. It can lead to increased apoptosis and inflammation-related gene expression levels, and is considered a reproductive toxicant that can cause testicular atrophy.

  • Appearance: Colorless to light yellow liquid
  • For research use only
  • Store at room temperature for 3 years
  • When in solvent, store at -80°C for 2 years or -20°C for 1 year

Catalog No. 50-004-52842


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.