Learn More
Medchemexpress LLC Dihexyl phthalate | 84-75-3 | 98.0% | 334.46 | 25 G
SDP
Supplier:  Medchemexpress LLC 25GHYW011215
Encompass_Preferred
Dihexyl phthalate is a commonly used phthalate ester found in various plastics and consumer products. It is categorized as a priority pollutant and an endocrine disruptor. This compound can lead to the accumulation of reactive oxygen species (ROS), promote inflammation, and significantly increase the expression levels of genes related to apoptosis and inflammation. Dihexyl phthalate is also known to cause testicular atrophy and is considered a reproductive toxicant.
  • Classified as a priority pollutant.
  • Identified as an endocrine disruptor.
  • Induces reactive oxygen species (ROS) accumulation.
  • Promotes inflammation.
  • Causes increased apoptosis and inflammation-related gene expression.
  • Can lead to testicular atrophy.
  • Considered a reproductive toxicant.
Catalog No. 50-004-51176