Learn More
Abcam Dihomo-gamma-linolenic acid (20:3, n-6), Long-chain polyunsaturated fatty acid, 100MG
Supplier:  Abcam AB145213100MG
Encompass_Preferred
MW 306.49 g/mol, Purity >99%. Long-chain polyunsaturated fatty acid. Cyclooxygenase and lipoxygenase substrate (IC₅₀ =45 μM, LTB4 formation). Biosynthetic PGE₁ precursor. Elongase product of γ-linolenic acid (ab145168). Arachidonic acid (ab120916) analog. Energy source in vivo. Orally active.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2077642