Learn More
Medchemexpress LLC Dihomo-r-Linolenic a 5mg | 21061-10-9 | 322.53 g·mol⁻¹ | C21H38O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1160155MG
Encompass_Preferred
Dihomo-γ-linolenic acid methyl ester is an n-6 polyunsaturated fatty acid methyl ester used as a research reagent. It is a metabolic precursor in the arachidonic acid pathway and is metabolized to anti-inflammatory eicosanoids such as prostaglandin E1. This highly purified compound is intended for biochemical and pharmacological studies investigating eicosanoid metabolism and inflammatory signaling.
  • High purity: ≥98%.
  • CAS number 21061-10-9.
  • Small-quantity research format (e.g., 5 MG aliquots in solvent).
  • Used for studies of eicosanoid metabolism and anti-inflammatory pathways.
  • Provided as a methyl ester derivative for lipid research workflows.
Catalog No. 50-002-27125