Learn More
Cayman Chemical DIhomoLInolenoyl Ethnolam 10mg
SDP
Supplier:  Cayman Chemical 9023510 MG
Encompass_Preferred
An endocannabinoid containing dihomo-γ-linoleate in place of the arachidonate moiety of AEA; binds to recombinant human CB1 and CB2 receptors with Ki values of 857 and 598 nM, respectively
Catalog No. 50-289-3415