Learn More
Medchemexpress LLC Dihydrexidine (hydrochloride) | 137417-08-4 | MFCD01860859 | 98.9% | 303.78 g/mol | C17H18ClNO2 | 1 ML

Supplier: Medchemexpress LLC HY101299B10MM 1ML
Dihydrexidine hydrochloride is a potent, selective full-efficacy D1/D5 dopamine receptor agonist used as a research reagent in pharmacology and neuroscience studies. The product is supplied as a ready-to-use 10 mM solution in DMSO (1 mL) suitable for in vitro and preclinical applications.
- Potent, selective full-efficacy D1/D5 dopamine receptor agonist.
- Ready-to-use 10 mM solution in DMSO, 1 mL vial.
- High purity (~98.9%) as verified by LCMS.
- CAS number 137417-08-4 and molecular formula C17H18ClNO2.
- Molecular weight 303.78 g/mol.
- Intended for research use; follow SDS for handling and storage.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.