Learn More
Apexbio Technology LLC Dihydrexidine hydrochloride 158704-02-0 5mg
SDP
Supplier:  Apexbio Technology LLC B50285
Encompass_Preferred
Dihydrexidine hydrochloride is a small-molecule agonist targeting the dopamine D1 receptor It is designed to selectively activate the D1 receptor subtype thereby modulating cyclic AMP-mediated signal transduction pathways Dihydrexidine hydrochloride exerts its biological activity primarily through dopaminergic D1 receptor stimulation It demonstrates receptor activation with an EC50 value of approximately 72 21 nM Based on these pharmacological properties dihydrexidine hydrochloride holds research potential in investigating dopaminergic neurotransmission mechanisms and evaluating dopamine receptor involvement in neural function as well as in biological screening assays for receptor-mediated functional effects in neuronal models
Catalog No. 50-000-88631