Learn More
Medchemexpress LLC Dihydro-β-erythroidine hydrobromide | 29734-68-7 | MFCD00153793 | >98.0% | 356.26 g·mol⁻¹ | C16H21NO3·HBr | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1076701MG
Encompass_Preferred
Dihydro-β-erythroidine hydrobromide is a competitive antagonist of neuronal nicotinic acetylcholine receptors with preferential activity at α4-containing subtypes. It is used as a pharmacological tool in electrophysiology and behavioral studies to investigate nicotinic signaling and nicotine-related effects. Supplied as the hydrobromide salt, it is suitable for both in vitro and in vivo research applications.
  • Selective, competitive antagonist of neuronal nAChRs.
  • Shows selectivity for α4β2 and α4β4 receptor subtypes.
  • Orally bioavailable and active in vivo.
  • High purity (≥98%) suitable for research use.
  • Provided as a stable hydrobromide salt for handling and formulation.
Catalog No. 50-002-42083