Learn More
TARGETMOL CHEMICALS INC Dihydro-N-Caffeoyltyramine 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN75721MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Dihydro-N-Caffeoyltyramine is a compound extracted from Lycii Cortex that has antioxidant and antifungal activities and can be used to study fungal infections. Purity 100%
Catalog No. 50-287-9631