Learn More
Selleck Chemical LLC Dihydroactinidiolide S3962-25mg
SDP
Supplier:  Selleck Chemical LLC S396225MG
Encompass_Preferred
Dihydroactinidiolide is known to be a bioactive molecule in animals It is a component of pheromones in insects such as red fire ants and in mammals such as the Cat and the Red Fox
Catalog No. 50-310-8120