Learn More
Cayman Chemical DIhydroartemIsInIc AcId 5mg
SDP
Supplier:  Cayman Chemical 251425 MG
Encompass_Preferred
A biosynthetic precursor to artemisinin; an intermediate in the synthesis of artemisinin from artemisinic acid
Catalog No. 50-293-5996