Learn More
TARGETMOL CHEMICALS INC Dihydrobaicalein 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN38481MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. Dihydrobaicalein is a natural product from Scutellaria baicalensis, a PLK1 inhibitor (IC506.3 uM).Dihydrobaicalein inhibits VRK2 and PLK2. Purity 99.86%
Catalog No. 50-288-0292