Learn More
Cayman Chemical DIhydrocaffeIc AcId 25g
SDP
Supplier:  Cayman Chemical 2839025 G
Encompass_Preferred
A polyphenol; scavenges ABTS (IC50 = 81.86 μg/ml) and DPPH (IC50 = 192.86 μg/ml) radicals; increases the survival of RGC-5 mouse retinal ganglion cells under hypoxic conditions and in the presence of SNAP in a concentration-dependent manner; decreases eNOS activity in EA.hy926 human endothelial cells in a concentration-dependent manner; decreases infarct size in a rat model of transient ischemia induced by MCAO at 30 mg/kg
Catalog No. 50-294-0511