Learn More
Cayman Chemical DIhydrochlamydocIn 250ug
SDP
Supplier:  Cayman Chemical 21614250 UG
Encompass_Preferred
A cyclic tetrapeptide analogue of the HDAC inhibitor chlamydocin; inhibits histone H4 peptide deacetylation in HeLa cells (IC50 = 30 nM); inhibits proliferation of A7r5 rat aortic smooth muscle cells at 1µM
Catalog No. 50-296-4257