Learn More
Medchemexpress LLC Dihydroconiferyl alcohol | 2305-13-7 | MFCD00016571 | 95.0% | 182.22 g/mol | C10H14O3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYW016393
Encompass_Preferred
Dihydroconiferyl alcohol (CAS 2305-13-7) is a plant-derived phenolic compound provided as a research reference standard. It is reported as a solid with 95% purity, molecular formula C10H14O3, and molecular weight 182.22 g/mol. The compound is used in analytical and biological studies, including investigations of cell division and plant biochemistry.
  • Reference standard for analytical and research use.
  • Reported purity 95% suitable for standard applications.
  • Molecular formula C10H14O3 and molecular weight 182.22 g/mol.
  • Used in studies of cell division and plant-derived phenolics.
  • Available in multiple pack sizes for laboratory experiments.
Catalog No. 50-003-02843