Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Dihydroconiferyl alcohol | 2305-13-7 | MFCD00016571 | 99.9% | 182.22 g/mol | C10H14O3 | 50 MG
SDP

Supplier:  Medchemexpress LLC HYW01639350MG

Encompass_Preferred

Dihydroconiferyl alcohol is a research-grade phenolic compound used in biochemical and plant biology studies as a cell division factor and analytical reference. It is supplied as a solid or as a solution for assay development, analytical reference, and biological research.

  • High purity: 99.9%.
  • Chemical formula: C10H14O3.
  • Molecular weight: 182.22 g/mol.
  • Available in multiple package sizes including 50 mg and solution format.
  • Suitable as a reference standard for analytical methods and assay development.
  • Accompanied by safety documentation for laboratory use.

Catalog No. 50-003-02695


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.