Learn More
Medchemexpress LLC Dihydrocoumarin | 119-84-6 | 148.16 | 100 G
SDP
Supplier:  Medchemexpress LLC HYN1926100G
Encompass_Preferred
Dihydrocoumarin is a compound found in *Melilotus officinalis*. It is a yeast Sir2p inhibitor and also inhibits human SIRT1 and SIRT2. It is suitable for laboratory research and includes the following features and benefits:
  • Inhibits yeast Sir2p
  • Inhibits human SIRT1 and SIRT2
  • Increases cytotoxicity in the TK6 cell line
  • Increases apoptosis in the TK6 cell line
  • Increases p53 lysine 373 and 382 acetylation
Catalog No. 50-004-15551