Learn More
Medchemexpress LLC β-Dihydroequilin-d4 | 350819-99-7 | 99.0% | 274.39 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY138112S21MG
Encompass_Preferred
β-Dihydroequilin-d4 is a deuterium labeled β-Dihydroequilin, primarily used as a laboratory chemical. This compound incorporates stable heavy isotopes of hydrogen, carbon, and other elements, which are utilized as tracers for quantitation during drug development processes. Deuteration has been shown to potentially influence the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled β-Dihydroequilin
  • For research in isotope-labeled compounds
  • Aids in drug quantitation during development
  • May impact drug pharmacokinetic and metabolic profiles
  • Ships at room temperature for less than two weeks
  • Store powder at -20°C for 3 years or 4°C for 2 years
  • Store in solvent at -80°C for 6 months or -20°C for 1 month
  • Appears as a solid
Catalog No. 50-003-47774