Learn More
Medchemexpress LLC Dihydroergotoxine (mesylate) | 8067-24-1 | 99.6% | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHYB0799
Encompass_Preferred
Dihydroergotoxine mesylate (Ergoloid mesylates) is an α-adrenergic blocking agent. It binds with high affinity to the GABAA receptor-associated Cl- channel and interacts with central dopaminergic and serotonergic receptors. It displays antiproliferative, antihypertensive, and neuroprotective activity.
  • Inhibits proliferation of human prostate cancer cell lines.
  • Stronger inhibitor of low-Km phosphodiesterase than high-Km phosphodiesterase.
  • Prevents delayed neuronal death in gerbil hippocampus.
  • Exerts a protective effect on oligemically disturbed brain metabolism.
  • Alters sleep-wakefulness cycle by increasing wakefulness.
  • Decreases blood pressure in spontaneously hypertensive rats.
Catalog No. 50-004-05723