Learn More
TARGETMOL CHEMICALS INC DIHYDROETHIDIUM 50MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T202150MG
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Dihydroethidium (PD-MY 003) is a superoxide indicator. purity: 99%
Catalog No. 50-257-7244