Learn More
Medchemexpress LLC Dihydrojasmone | 1128-08-1 | 1 ML

Supplier: Medchemexpress LLC HYN709810MM/1ML
Dihydrojasmone, a constituent of bergamot oil, is an important perfume ingredient. This product is for research use only.
- Purity: 98.35%
- Molecular weight: 166.26
- Formula: C11H18O
- Appearance: Liquid (density: 0.916 g/cm³)
- Color: Colorless to light yellow
- Storage (pure form): -20°C for 3 years, 4°C for 2 years
- Storage (in solvent): -80°C for 6 months, -20°C for 1 month
- Solubility in vitro: DMSO : 100 mg/mL (601.47 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.