Learn More
Medchemexpress LLC Dihydrojasmone | 1128-08-1 | 10 G

Supplier: Medchemexpress LLC HYN709810G
Dihydrojasmone is a constituent of bergamot oil and is an important perfume ingredient. This product is intended for research use only and is not sold to patients.
- High purity: 98.35%
- Colorless to light yellow liquid appearance
- Molecular Weight: 166.26
- Chemical formula: C11H18O
- Shipping at room temperature in continental US
- Storage conditions vary for pure form and in solvent
- Soluble in DMSO and various in vivo protocols
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.