Learn More
Medchemexpress LLC Dihydrojasmone | 1128-08-1 | 5 G

Supplier: Medchemexpress LLC HYN70985G
Dihydrojasmone, a constituent of bergamot oil, is an important perfume ingredient. This product is for research use only. It appears as a colorless to light yellow liquid with a purity of 98.35%.
- Molecular weight: 166.26
- Formula: C11H18O
- Appearance: Colorless to light yellow liquid
- Density: 0.916 g/cm3
- Structure classification: Ketones, aldehydes, acids
- Initial source: Plants other families
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.