Learn More
Medchemexpress LLC Dihydrojasmone | 1128-08-1 | MFCD00036480 | 25 G

Supplier: Medchemexpress LLC 25GHYN7098
Dihydrojasmone is a constituent of bergamot oil and is an important perfume ingredient. It is for research use only.
- Constituent of bergamot oil
- Important perfume ingredient
- Purity: 98.35%
- Appearance: Liquid (density: 0.916 g/cm3)
- Color: Colorless to light yellow
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.