Learn More
Medchemexpress LLC Dihydrojasmonic acid | 3572-64-3 | 99.3% | 100 MG

Supplier: Medchemexpress LLC HY131116100MG
Dihydrojasmonic acid is a plant growth regulator.
- Molecular weight: 212.29
- Chemical formula: C12H20O3
- Appears as a colorless to light yellow liquid with a density of 1.038±0.06 g/cm3
- Pure form can be stored at -20°C for 3 years or 4°C for 2 years
- When in solvent, store at -80°C for 6 months or -20°C for 1 month
- Solubility in DMSO: 100 mg/mL (471.05 mM)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.