Learn More
Medchemexpress LLC Dihydrokavain | 587-63-3 | 232.28 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN092010MM/1ML
Encompass_Preferred
Dihydrokavain (7,8-Dihydrokawain) is a natural kavalactone compound. It functions as an inhibitor of COX-1, COX-2, CYP2C9, CYP2C19, and CYP3A4. This compound has been shown to reduce TNFα secretion and exhibit analgesic and anxiolytic effects.
  • Natural kavalactone compound
  • Inhibits COX-1 and COX-2
  • Inhibits CYP2C9, CYP2C19, and CYP3A4
  • Reduces TNFα secretion
  • Demonstrates analgesic and anxiolytic effects
  • Purity: 99.94%
  • For research use only
Catalog No. 50-004-16835