Learn More
Medchemexpress LLC Dihydrolanosterol | 79-62-9 | 99.6% | 1 MG

Supplier: Medchemexpress LLC HY1224101MG
Dihydrolanosterol is a sterol that serves as a CYP51 substrate and a cholesterol biosynthesis inhibitor. It is supplied for research use only as a white to off-white solid in small milligram quantities.
- acts as a substrate of CYP51 for studies of sterol metabolism
- high purity suitable for analytical and biochemical assays
- molecular formula C30H52O, molecular weight 428.73 g/mol
- available in milligram pack sizes for small-scale experiments
- store at 4°C and protect from light; in solution store at -80°C for long-term stability
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.