Learn More
Medchemexpress LLC Dihydrolapachenole | 20213-26-7 | 5 MG

Supplier: Medchemexpress LLC HYN76385MG
Dihydrolapachenole is a laboratory chemical primarily used in the manufacture of various substances. This solid compound has a molecular formula of C16H18O2 and a molecular weight of 242.31.
- Laboratory chemical application
- Used for substance manufacturing
- Solid form
- Molecular formula: C16H18O2
- Molecular weight: 242.31
- Causes skin irritation (Category 2)
- Causes serious eye irritation (Category 2A)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.