Learn More
Medchemexpress LLC HY-N2403 5mg Medchemexpress, Dihydrolycorine CAS:6271-21-2 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN24035MG
Encompass_Preferred
Medchemexpress, HY-N2403 5mg Dihydrolycorine CAS:6271-21-2 Dihydrolycorine, isolated from Lycoris radiate Herb, is an inhibitor of protein synthesis in eukarytic cells by inhibiting the peptide bone formation step. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2893