Learn More
Medchemexpress LLC Dihydromunduletone | 674786-20-0 | 99.2% | 424.49 | C25H28O6 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY101483100MG
Encompass_Preferred
Dihydromunduletone is a rotenoid-derived small molecule used as an analytical research standard and described as a selective antagonist of adhesion G protein-coupled receptors. It is supplied as a light yellow solid with a reported purity of 99.23%, molecular formula C25H28O6, and molecular weight 424.49, intended for in vitro research and analytical applications.
  • Selective antagonist of adhesion G protein-coupled receptors.
  • High reported purity (99.23%) suitable for analytical work.
  • Light yellow solid form for convenient handling and weighing.
  • Recommended storage conditions provided to preserve stability.
  • Available in research-scale quantities, including 100 mg.
Catalog No. 50-002-46928