Learn More
Medchemexpress LLC HY-N4240 5mg Medchemexpress, Dihydropalmatine CAS:26067-60-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN42405MG
Encompass_Preferred
Medchemexpress, HY-N4240 5mg Dihydropalmatine CAS:26067-60-7 Dihydropalmatine is a alkaloid isolated from Berberis aristata. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2928