Learn More
Medchemexpress LLC Dihydrotanshinone I | 87205-99-0 | 278.30 | 5 MG

Supplier: Medchemexpress LLC HYN03605MG
Dihydrotanshinone I is a natural compound extracted from *Salvia miltiorrhiza Bunge*. This compound has been extensively used for treating cardiovascular diseases and demonstrates an entry-blocking effect against MERS-CoV.
- Extracted from *Salvia miltiorrhiza Bunge*
- Used for treating cardiovascular diseases
- Exhibits entry-blocking effect for MERS-CoV
- Attenuates atherosclerosis in ApoE-/- mice
- Inhibits LOX-1, NOX4, and NF-κB expression in aorta
- Improves cardiac function and reduces infarct size in myocardial ischemia-reperfusion rats
- Induces caspase-dependent apoptosis and autophagy in colon cancer cells
- Potentially a safe, high-throughput screening compound for MERS-CoV entry inhibitors
- Purity: 99.57%
- Appearance: Solid, brown to red
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.