Learn More
Medchemexpress LLC Dihydrotetrabenazine | 3466-75-9 | 99.2% | C19H29NO3 | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY141554
Encompass_Preferred
Dihydrotetrabenazine (DHTBZ) is the active metabolite of Tetrabenazine. It functions as an inhibitor for human vesicular monoamine transporter 2 (VMAT2), which results in a reduction of monoamine content in presynaptic neurons. This compound can be used in movement disorder research.
  • Active metabolite of tetrabenazine
  • Functions as a VMAT2 inhibitor
  • Reduces monoamine content in presynaptic neurons
  • Suitable for movement disorder research
  • Appearance: solid
  • Color: white to off-white
Catalog No. 50-003-52557