Learn More
Medchemexpress LLC Dihydrouracil-d4 | 334473-41-5 | 99.97% | 118.13 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYW012926S11MG
Encompass_Preferred
Dihydrouracil-d4 is the deuterium-labeled form of Dihydrouracil, a metabolite of Uracil. It can be used as a marker for identifying individuals with dihydropyrimidine dehydrogenase (DPD)-deficiency. This compound is intended for research use only, serving as a tracer and internal standard for quantitative analysis. Deuteration has gained attention for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis via NMR, GC-MS, or LC-MS.
  • Serves as a marker for identifying individuals with dihydropyrimidine dehydrogenase (DPD)-deficiency.
  • Intended for research use only.
  • Deuteration can impact pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54134