Learn More
Medchemexpress LLC Dihydrouracil-d4 | 334473-41-5 | 99.97% | 118.13 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYW012926S15MG
Encompass_Preferred
Dihydrouracil-d4 is the deuterium labeled Dihydrouracil. Dihydrouracil is a metabolite of Uracil and can be used as a marker for identification of dihydropyrimidine dehydrogenase (DPD)-deficient individuals. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Stable heavy isotopes of hydrogen, carbon, and other elements.
  • Incorporated into drug molecules for tracer and quantitation during drug development.
  • Deuteration has potential to affect pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-54080