Learn More
Cayman Chemical OctnoylCoenzyme AsodIum s 25mg
SDP
Supplier:  Cayman Chemical 2786825 MG
Encompass_Preferred
A medium-chain acyl CoA; levels increased in liver in patients with Reye’s syndrome and β-oxidation of octanoyl-CoA by MCADH is decreased in patients with MCD; inhibits citrate synthase and glutamate dehydrogenase
Catalog No. 50-294-1755