Learn More
Medchemexpress LLC DiI | 41085-99-8 | 99.98% | 100 MG
SDP
Supplier:  Medchemexpress LLC HYD0083100MG
Encompass_Preferred
DiI is a long-chain carbocyanine dye. This lipophilic carbocyanine fluorescent dye exhibits red fluorescence and is suitable for cell membrane and nuclei labeling, long-term monitoring of cells, immunofluorescence staining of exosomes, and exosome uptake assays. It is for research use only.
  • Used to label cells, organelles, liposomes, viruses, and lipoproteins.
  • Ideal for staining cytoplasmic membranes of cells.
  • Diffuses laterally within the plasma membrane, staining the entire cell.
  • Exhibits distinct orange fluorescence, facilitating multicolor imaging and flow cytometric analysis of live cells.
  • Can be used with standard TRITC filters.
  • Exhibits very low cell toxicity.
  • Widely used for determining lipoproteins such as LDL and HDL.
  • Labeled motoneurons have remained viable for up to 4 weeks in culture and up to one year in vivo.
Catalog No. 50-004-10663