Learn More
Medchemexpress LLC DiI | 41085-99-8 | 99.98% | 5 MG
SDP
Supplier:  Medchemexpress LLC HYD00835MG
Encompass_Preferred
DiI is a long-chain carbocyanine dye frequently used to label cells, organelles, liposomes, viruses, and lipoproteins. This lipophilic fluorescent dye exhibits red fluorescence and is particularly effective for labeling cell membranes. Its very low cell toxicity makes it suitable for various biological applications, including the determination of lipoproteins such as LDL and HDL.
  • Long-chain carbocyanine dye
  • Labels cells, organelles, liposomes, viruses, and lipoproteins
  • Lipophilic fluorescent dye
  • Exhibits red fluorescence
  • Can label cell membranes and liposomes
  • Very low cell toxicity
  • Used for determining lipoproteins
Catalog No. 50-004-10034