Learn More
Selleck Chemical LLC DiI E4783-25MG
SDP
Supplier:  Selleck Chemical LLC E478325MG
Encompass_Preferred
DiI (DiIC18(3)) is a lipid-soluble fluorescent dye that traces neuronal pathways through retrograde and anterograde transport in neurons It labels cell bodies axons dendrites and dendritic spines offering high brightness slow fading and long-lasting persistence without leakage It is employed to investigate brain connectivity including in postmortem human brain tissue
Catalog No. 50-000-42000