Learn More
Aat Bioquest DiIC123 perchlorate [11-Didodecyl-3333-tetramethylindocarbocyanine perchlorate] (25 mg)
SDP
Supplier:  Aat Bioquest 22035
Encompass_Preferred
DiI DiO DiD and DiR dyes are a family of lipophilic fluorescent stains for labeling membranes and other hydrophobic structures. This catalog is for 25 mg.
Catalog No. 50-225-0032