Learn More
Medchemexpress LLC Diiodo(p-cymene)ruthenium(II) dimer | 90614-07-6 | C20H28I4Ru2 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY79986100MG
Encompass_Preferred
Diiodo(p-cymene)ruthenium(II) dimer is classified as harmful if swallowed (H302) and causes serious eye irritation (H319). It is a solid with a melting/freezing point of 234-236 °C. The product is for research use only and should be handled by suitably qualified experienced scientists in appropriately equipped and authorized facilities.
  • Harmful if swallowed
  • Causes serious eye irritation
  • Solid appearance
  • Melting/freezing point of 234-236 °C
  • Stable under recommended storage conditions
  • Recommended storage at 4°C, protected from light, and under nitrogen
  • If in solvent, store at -80°C for 6 months or -20°C for 1 month (protected from light, stored under nitrogen)
  • Shipping at room temperature if less than 2 weeks
Catalog No. 50-004-33287